accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19395-41-6

IUPAC Name: 2-phenyl-2-(piperidin-2-yl)acetic acid
Synonyms: ritalinic acid
Molecular Weight (g/mol): 219.28
Molecular Formula: C13H17NO2
InChi Key: INGSNVSERUZOAK-UHFFFAOYNA-N
SMILES: OC(=O)C(C1CCCCN1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)