accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 193978-23-3

IUPAC Name: 4,4,5,5-tetramethyl-2-(thiophen-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(thiophen-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 210.1
Molecular Formula: C10H15BO2S
InChi Key: FFZHICFAHSDFKZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CS1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)