accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19398-06-2

IUPAC Name: (2-ethylphenyl)hydrazine
Synonyms: (2-ethylphenyl)hydrazine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: JHPOWXCLWLEKBY-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1NN
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results