accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3CH3

CAS RN 19398-77-7

IUPAC Name: 3,4-diethylhexane
Synonyms: hexane, 3,4-diethyl-
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: VBZCRMTUDYIWIH-UHFFFAOYSA-N
SMILES: CCC(CC)C(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results