accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOO

CAS RN 194017-69-1

IUPAC Name: 4-(pyridin-2-yloxy)benzaldehyde
Synonyms: 4-(pyridin-2-yloxy)benzaldehyde
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: DPRZACGKYIDYCK-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OC2=CC=CC=N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results