accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 19402-87-0

IUPAC Name: 9-benzyl-9H-carbazole
Synonyms: 9-benzylcarbazole
Molecular Weight (g/mol): 257.34
Molecular Formula: C19H15N
InChi Key: HBAKJBGOHINNQM-UHFFFAOYSA-N
SMILES: C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results