accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3H3CONOO

CAS RN 1941-24-8

IUPAC Name: tetramethylazanium nitrate
Synonyms: tetramethylammonium nitrate
Molecular Weight (g/mol): 136.15
Molecular Formula: C4H12N2O3
InChi Key: QGPVYHSXZFOSRL-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O.C[N+](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results