accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 1942-46-7

IUPAC Name: dec-5-yne
Synonyms: 5-decyne
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: JWBQJUFCNOLNNC-UHFFFAOYSA-N
SMILES: CCCCC#CCCCC
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)