accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNH3CSH

CAS RN 1942-52-5

IUPAC Name: hydrogen 2-(diethylamino)ethane-1-thiol chloride
Synonyms: hydrogen ethanethiol, 2-(diethylamino)- chloride
Molecular Weight (g/mol): 169.71
Molecular Formula: C6H16ClNS
InChi Key: QAXLTZBBDSDRPW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results