accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1943-11-9

IUPAC Name: trimethyl(nonyl)azanium bromide
Synonyms: trimethyl(nonyl)azanium bromide
Molecular Weight (g/mol): 266.27
Molecular Formula: C12H28BrN
InChi Key: SSEGNMJSEROZIF-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results