accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1943-82-4

IUPAC Name: (2-isocyanatoethyl)benzene
Synonyms: phenylethyl isocyanate
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: HACRKYQRZABURO-UHFFFAOYSA-N
SMILES: O=C=NCCC1=CC=CC=C1
Boiling Point 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)