accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1943-83-5

IUPAC Name: 1-chloro-2-isocyanatoethane
Synonyms: 1-chloro-2-isocyanatoethane
Molecular Weight (g/mol): 105.52
Molecular Formula: C3H4ClNO
InChi Key: BCMYXYHEMGPZJN-UHFFFAOYSA-N
SMILES: ClCCN=C=O
Boiling Point 
  • (7)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (7)
  • (1)
  • (4)
  • (4)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)