accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2N

CAS RN 19430-83-2

IUPAC Name: 4-[(3-aminophenyl)methyl]aniline
Synonyms: 4-[(3-aminophenyl)methyl]aniline
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: FGWQCROGAHMWSU-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=CC(N)=C2)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results