accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OClCl

CAS RN 19433-84-2

IUPAC Name: 2,3-dichloropropanamide
Synonyms: propanamide, 2,3-dichloro-
Molecular Weight (g/mol): 141.98
Molecular Formula: C3H5Cl2NO
InChi Key: SQGUQSRDWXDLIH-UHFFFAOYNA-N
SMILES: NC(=O)C(Cl)CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results