accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19434-34-5

IUPAC Name: 2-phenoxybenzaldehyde
Synonyms: 2-phenoxybenzaldehyde
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: IMPIIVKYTNMBCD-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1OC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results