accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19437-26-4

IUPAC Name: 2-(pyridine-2-carbonyl)pyridine
Synonyms: 2-(pyridine-2-carbonyl)pyridine
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: QPOWUYJWCJRLEE-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=N1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (4)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)