accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19438-61-0

IUPAC Name: 5-methyl-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-methyl-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: ZOXBWJMCXHTKNU-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(=O)OC(=O)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)