accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 194416-45-0

IUPAC Name: N-(4-bromophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
Synonyms: N-(4-bromophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
Molecular Weight (g/mol): 384.27
Molecular Formula: C20H18BrNO2
InChi Key: XXCDCFFPMMCPEE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results