accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 194421-56-2

IUPAC Name: 6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
Synonyms: 6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: DRMNZTFLOOSXIN-UHFFFAOYNA-N
SMILES: CC1(C)C2C1C(=O)NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results