CAS RN 194423-15-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PD 168393
A potent, cell-permeable, irreversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50 = 700 pM).