accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OCH3CH3OCH3CH3OLa

CAS RN 19446-52-7

IUPAC Name: lanthanum(3+) tris(propan-2-olate)
Synonyms: lanthanum(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 316.17
Molecular Formula: C9H21LaO3
InChi Key: SORGMJIXNUWMMR-UHFFFAOYSA-N
SMILES: [La+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)