CAS RN 19455-20-0
IUPAC Name:
potassium 2-methylpropanoate
Synonyms:
potassium isobutyrate
Molecular Weight (g/mol):
126.2
Molecular Formula:
C4H7KO2
InChi Key:
LBOHISOWGKIIKX-UHFFFAOYSA-M
SMILES:
[K+].CC(C)C([O-])=O
1
–
1
of
1
results
1
Potassium Isobutyrate 98.0+%, TCI America™
CAS: 19455-20-0 Molecular Formula: C4H7KO2 Molecular Weight (g/mol): 126.196 MDL Number: MFCD00058992 InChI Key: LBOHISOWGKIIKX-UHFFFAOYSA-M Synonym: Isobutyric Acid Potassium Salt PubChem CID: 23677462 IUPAC Name: potassium;2-methylpropanoate SMILES: CC(C)C(=O)[O-].[K+]