accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHIOI

CAS RN 1948-40-9

IUPAC Name: 4-hydroxy-3,5-diiodobenzaldehyde
Synonyms: 4-hydroxy-3,5-diiodobenzaldehyde
Molecular Weight (g/mol): 373.92
Molecular Formula: C7H4I2O2
InChi Key: WHLUEIMENHLCMY-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(C=O)C=C1I
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)