accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrN

CAS RN 19481-82-4

IUPAC Name: 2-bromopropanenitrile
Synonyms: 2-bromopropanenitrile
Molecular Weight (g/mol): 133.98
Molecular Formula: C3H4BrN
InChi Key: PYNYHMRMZOGVML-UHFFFAOYNA-N
SMILES: CC(Br)C#N
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results