accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClO

CAS RN 19484-57-2

IUPAC Name: 6-chloro-2-hydroxy-4H-chromen-4-one
Synonyms: 6-chloro-2-hydroxychromen-4-one
Molecular Weight (g/mol): 196.59
Molecular Formula: C9H5ClO3
InChi Key: UJHIADILZZWIED-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C2=CC(Cl)=CC=C2O1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results