CAS RN 194918-76-8
IUPAC Name:
4-aminocyclopentane-1,2,4-tricarboxylic acid
Synonyms:
4-aminocyclopentane-1,2,4-tricarboxylic acid
Molecular Weight (g/mol):
217.18
Molecular Formula:
C8H11NO6
InChi Key:
FERIKTBTNCSGJS-UHFFFAOYNA-N
SMILES:
NC1(CC(C(C1)C(O)=O)C(O)=O)C(O)=O
1
–
1
of
1
results
1
ACPT-I, Tocris Bioscience™
CAS: 194918-76-8 Molecular Formula: C8H11NO6 Molecular Weight (g/mol): 217.177 InChI Key: FERIKTBTNCSGJS-OBLUMXEWSA-N Synonym: acpt-i,acpt-ii,1-aminocyclopentane-1,3,4-tricarboxylic acid,1r,2s-4-aminocyclopentane-1,2,4-tricarboxylic acid,4-amino-cyclopentane-1,2,4-tricarboxylic acid,tocris-1113,d0w3py,1s,3r,4s-1-aminocyclopentane-1,3,4-tricarboxylic acid,1r,2s,4s-4-aminocyclopentane-1,2,4-tricarboxylic acid,1r,3r,4s-1-aminocyclopentane-1,3,4-tricarboxylic acid PubChem CID: 6604820 IUPAC Name: (1R,2S)-4-aminocyclopentane-1,2,4-tricarboxylic acid SMILES: C1C(C(CC1(C(=O)O)N)C(=O)O)C(=O)O