accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHONOO

CAS RN 19499-93-5

IUPAC Name: 2,3-dimethyl-4-nitrophenol
Synonyms: 2,3-dimethyl-4-nitrophenol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DAEZHJNJHQAEPE-UHFFFAOYSA-N
SMILES: CC1=C(O)C=CC(=C1C)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)