CAS RN 194992-44-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PESTANAL™ Acetochlor OA Analytical Standard, MilliporeSigma™ Supelco™
Acetochlor OA is a metabolite of the preemergence herbicide, acetochlor. It is used to control annual grasses and broad-leaved weeds in corn, soybeans, and other crops.