accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19501-58-7

IUPAC Name: (4-methoxyphenyl)hydrazine hydrochloridyl
Synonyms: (4-methoxyphenyl)hydrazine hydrochloridyl
Molecular Weight (g/mol): 173.62
Molecular Formula: C7H10ClN2O
InChi Key: UNLQTHFXAIISEP-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=C(NN)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)