accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CH3C

CAS RN 195062-59-0

IUPAC Name: 4,4,5,5-tetramethyl-2-(2-methylphenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2-methylphenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 218.1
Molecular Formula: C13H19BO2
InChi Key: NUFSJKMRPYGNHV-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results