accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COBOH3CH3CCl

CAS RN 195062-61-4

IUPAC Name: 2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 238.52
Molecular Formula: C12H16BClO2
InChi Key: NYARTXMDWRAVIX-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (2)