accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBOH3CH3CH3CCH3O

CAS RN 195062-62-5

IUPAC Name: ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 276.14
Molecular Formula: C15H21BO4
InChi Key: NCVIYKCFTYSAGN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results