accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19513-05-4

IUPAC Name: manganese(3+) triacetate
Synonyms: manganese(III) acetate
Molecular Weight (g/mol): 232.07
Molecular Formula: C6H9MnO6
InChi Key: AHSBSUVHXDIAEY-UHFFFAOYSA-K
SMILES: [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)