accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OOHOOH2N

CAS RN 195189-45-8

IUPAC Name: 3,5-bis(4-aminophenoxy)benzoic acid
Synonyms: 3,5-bis(4-aminophenoxy)benzoic acid
Molecular Weight (g/mol): 336.35
Molecular Formula: C19H16N2O4
InChi Key: KPKOSOUTWDOOIW-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC(=CC(OC3=CC=C(N)C=C3)=C2)C(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results