accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 195191-47-0

IUPAC Name: 2-bromo-4-chloro-6-fluoroaniline
Synonyms: 2-bromo-4-chloro-6-fluoroaniline
Molecular Weight (g/mol): 224.46
Molecular Formula: C6H4BrClFN
InChi Key: HBHBARSMRVAINH-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(Cl)C=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results