accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOHOHHO

CAS RN 195385-93-4

IUPAC Name: 7-{[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-4-methyl-2H-chromen-2-one
Synonyms: 7-{[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-4-methylchromen-2-one
Molecular Weight (g/mol): 308.29
Molecular Formula: C15H16O7
InChi Key: FAGLTVBWEMHJRP-UHFFFAOYNA-N
SMILES: CC1=CC(=O)OC2=CC(OC3OC(CO)C(O)C3O)=CC=C12
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)