accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19541-95-8

IUPAC Name: 5-(4-methoxyphenyl)-1H-pyrazol-3-amine
Synonyms: 5-(4-methoxyphenyl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: UPAGEJODHNVJNM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC(N)=NN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)