accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 195443-34-6

IUPAC Name: 4'-iodo-N-(3-methylphenyl)-N-phenyl-[1,1'-biphenyl]-4-amine
Synonyms: 4'-iodo-N-(3-methylphenyl)-N-phenyl-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 461.35
Molecular Formula: C25H20IN
InChi Key: AKQZSFMSAULQQN-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(I)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results