accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3HO

CAS RN 19550-03-9

IUPAC Name: 2,3-dimethylhexan-2-ol
Synonyms: 2-hexanol, 2,3-dimethyl-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: BFKOEFFCVFMWPF-UHFFFAOYNA-N
SMILES: CCCC(C)C(C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results