accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19550-07-3

IUPAC Name: 2,5-dimethylhexan-3-ol
Synonyms: 3-hexanol, 2,5-dimethyl-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: SNKTZHPOKPYBPT-UHFFFAOYNA-N
SMILES: CC(C)CC(O)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results