accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19550-56-2

IUPAC Name: 2,9-dimethyldec-5-yne
Synonyms: 2,9-dimethyldec-5-yne
Molecular Weight (g/mol): 166.31
Molecular Formula: C12H22
InChi Key: RCFYJIGZQAOTFV-UHFFFAOYSA-N
SMILES: CC(C)CCC#CCCC(C)C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)