accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19550-57-3

IUPAC Name: 6,7-dimethyl-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6,7-dimethyl-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: CVKLXAQUJMXVMC-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=C1C)C(=O)CCC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results