accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19550-75-5

IUPAC Name: (3E)-2,2-dimethylhept-3-ene
Synonyms: (3E)-2,2-dimethylhept-3-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: BQOCYCICSYUPRF-BQYQJAHWSA-N
SMILES: CCC\C=C\C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results