accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19552-10-4

IUPAC Name: (4-bromophenyl)methanethiol
Synonyms: (4-bromophenyl)methanethiol
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: CUCKXDPCCYHFMQ-UHFFFAOYSA-N
SMILES: SCC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results