CAS RN 1956-11-2
IUPAC Name:
4-nitrophenyl dodecanoate
Synonyms:
4-nitrophenyl dodecanoate
Molecular Weight (g/mol):
321.42
Molecular Formula:
C18H27NO4
InChi Key:
YNGNVZFHHJEZKD-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-Nitrophenyl Laurate 98.0+%, TCI America™
CAS: 1956-11-2 Molecular Formula: C18H27NO4 Molecular Weight (g/mol): 321.42 MDL Number: MFCD00047730 InChI Key: YNGNVZFHHJEZKD-UHFFFAOYSA-N Synonym: Lauric Acid 4-Nitrophenyl Ester PubChem CID: 74778 IUPAC Name: (4-nitrophenyl) dodecanoate SMILES: CCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]