CAS RN 195604-39-8
IUPAC Name:
1-cyclopropylethan-1-amine
Synonyms:
1-cyclopropylethanamine
Molecular Weight (g/mol):
85.15
Molecular Formula:
C5H11N
InChi Key:
IXCXVGWKYIDNOS-UHFFFAOYNA-N
SMILES:
CC(N)C1CC1
1
–
1
of
1
results
1
Promotions Available
(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%
(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%
CAS: 195604-39-8 Molecular Formula: C5H11N Molecular Weight (g/mol): 85.15 MDL Number: MFCD08064289 InChI Key: IXCXVGWKYIDNOS-BYPYZUCNSA-N PubChem CID: 7363995 IUPAC Name: (1S)-1-cyclopropylethanamine SMILES: CC(C1CC1)N