accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFFI

CAS RN 195624-84-1

IUPAC Name: 2-iodo-1-methoxy-4-(trifluoromethyl)benzene
Synonyms: 2-iodo-1-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 302.04
Molecular Formula: C8H6F3IO
InChi Key: UMUJZUDPHBABQX-UHFFFAOYSA-N
SMILES: COC1=C(I)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results