accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19567-78-3

IUPAC Name: disodium hexachloroiridiumdiuide hexahydrate
Synonyms: disodium hexachloroiridate(2-) hexahydrate
Molecular Weight (g/mol): 558.99
Molecular Formula: Cl6H12IrNa2O6
InChi Key: QZVJPVJIAOOTJU-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.[Na+].[Na+].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results