accessibility menu, dialog, popup

UserName

Viewing profile as user:
PtClClClClClClNaNaH2OH2OH2OH2OH2OH2O

CAS RN 19583-77-8

IUPAC Name: disodium hexachloroplatinum hexahydrate
Synonyms: disodium hexachloroplatinum hexahydrate
Molecular Weight (g/mol): 561.85
Molecular Formula: Cl6H12Na2O6Pt
InChi Key: XGFSWVNCHVSCSP-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
Grade 
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)