accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFF

CAS RN 195886-79-4

IUPAC Name: 2-(difluoromethyl)-1,4-difluorobenzene
Synonyms: 2-(difluoromethyl)-1,4-difluorobenzene
Molecular Weight (g/mol): 164.1
Molecular Formula: C7H4F4
InChi Key: FDUVCOVVYFCQEH-UHFFFAOYSA-N
SMILES: FC(F)C1=C(F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results